Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olfcq19
Entrez Gene ID
100003908
Gene Name
olfactory receptor C family, q19
Gene Aliases
si:dkey-10i19.2
Chromosome Location
Chr.18: 33818504 - 33823205 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083877.1 NP_001077346.1 4 - 5 1575 86
XM_009293692.2 XP_009291967.1 4 - 5 1579 86

View Full Product Details