Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
akap1b
Entrez Gene ID
556208
Gene Name
A kinase (PRKA) anchor protein 1b
Gene Aliases
akap1, fa08b04, wu:fa08b04
Chromosome Location
Chr.15: 30840393 - 30857350 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098179.2 NP_001091649.2 5 - 6 2099 66
XM_017350962.2 XP_017206451.1 5 - 6 2106 66
GenBank mRNA BC139889.1 - 6 - 7 1758 66

View Full Product Details