Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adcy2b
Entrez Gene ID
557902
Gene Name
adenylate cyclase 2b (brain)
Gene Aliases
si:dkey-266i6.1
Chromosome Location
Chr.19: 27609618 - 27776856 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099987.1 NP_001093457.1 11 - 12 1752 123

View Full Product Details