Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ryr1b
Entrez Gene ID
570245
Gene Name
ryanodine receptor 1b (skeletal)
Gene Aliases
im:7137738, ryrab, si:dkey-250i19.1
Chromosome Location
Chr.18: 35489483 - 35736799 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102571.1 NP_001096041.1 79 - 80 11250 138
XM_017351957.2 XP_017207446.1 80 - 81 11479 138
XM_017351958.2 XP_017207447.1 79 - 80 11464 138
XM_017351960.2 XP_017207449.1 80 - 81 11479 138
XM_021467693.1 XP_021323368.1 80 - 81 11479 138
XM_021467695.1 XP_021323370.1 79 - 80 11458 138
GenBank mRNA AB247454.1 - 79 - 80 11250 138

View Full Product Details