Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
anos1a
Entrez Gene ID
30630
Gene Name
anosmin 1a
Gene Aliases
anosmin-1a, kal1.1, kal1a, wu:fl91z45
Chromosome Location
Chr.1: 31943017 - 31979514 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131358.1 NP_571433.1 13 - 14 1957 80
GenBank mRNA AF163310.1 - 13 - 14 1957 80
BC060938.1 - 13 - 14 1961 80
KM655630.1 - 13 - 14 1910 80

View Full Product Details