Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nots
Entrez Gene ID
114367
Gene Name
nothepsin
Gene Aliases
LAP, ctsel, zgc:103608
Chromosome Location
Chr.4: 78010696 - 78020284 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131804.1 NP_571879.1 - 67 66
GenBank mRNA AJ278269.1 - - 67 66
BC086835.1 - - 232 66
BC165335.1 - - 182 66

View Full Product Details