Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ldlrap1a
Entrez Gene ID
368278
Gene Name
low density lipoprotein receptor adaptor protein 1a
Gene Aliases
ldlrap1, sb:cb50, zgc:56121
Chromosome Location
Chr.19: 29337774 - 29363780 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198980.1 NP_945331.1 5 - 6 613 67
XM_005159645.3 XP_005159702.1 5 - 6 665 67
GenBank mRNA BC045926.1 - 5 - 6 613 67
BC164648.1 - 5 - 6 558 67

View Full Product Details