Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ercc4
Entrez Gene ID
327322
Gene Name
excision repair cross-complementation group 4
Gene Aliases
fancq, fi03a05, wu:fi03a05, zgc:63468
Chromosome Location
Chr.12: 19959037 - 19974532 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199785.1 NP_956079.1 4 - 5 757 72
GenBank mRNA BC054895.1 - 4 - 5 757 72

View Full Product Details