The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200950.1 | NP_957244.1 | 5 - 6 | 527 | 83 |
GenBank mRNA | BC053162.1 | - | 5 - 6 | 527 | 83 |