Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rtn4rl2a
Entrez Gene ID
403307
Gene Name
reticulon 4 receptor-like 2 a
Gene Aliases
NgRH1a
Chromosome Location
Chr.1: 44491077 - 44493548 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203479.1 NP_982346.1 2 - 3 385 90
GenBank mRNA AY263332.1 - 2 - 3 385 90
BC162356.1 - 2 - 3 306 90
BC162359.1 - 2 - 3 306 90
CU468805.1 - 2 - 3 282 90

View Full Product Details