Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmo4b
Entrez Gene ID
324849
Gene Name
LIM domain only 4b
Gene Aliases
fc45c09, lmo4l, wu:fc45c09
Chromosome Location
Chr.6: 35161 - 54400 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212689.2 NP_997854.2 3 - 4 705 60
GenBank mRNA AF398515.2 - 3 - 4 661 60
BC076053.1 - 3 - 4 679 60

View Full Product Details