Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prg4a
Entrez Gene ID
336351
Gene Name
proteoglycan 4a
Gene Aliases
CH1073-202I11.1, fb08a02, wu:fb08a02, wu:fk21a06, zgc:56698
Chromosome Location
Chr.2: 20793955 - 20812063 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212753.2 NP_997918.2 2 - 3 154 84
XM_005163364.4 XP_005163421.1 2 - 3 177 84
GenBank mRNA BC050178.1 - 2 - 3 154 84

View Full Product Details