Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aip
Entrez Gene ID
334795
Gene Name
aryl hydrocarbon receptor interacting protein
Gene Aliases
wu:fa03h10, zgc:101101
Chromosome Location
Chr.1: 39995074 - 40009755 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214712.1 NP_999877.1 1 - 2 98 107
GenBank mRNA AF274877.1 - 1 - 2 98 107
BC078231.1 - 1 - 2 122 107

View Full Product Details