Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2x8
Entrez Gene ID
431734
Gene Name
cytochrome P450, family 2, subfamily X, polypeptide 8
Gene Aliases
zgc:192870, zgc:91876
Chromosome Location
Chr.25: 17388661 - 17395315 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002187.1 NP_001002187.1 3 - 4 502 103
GenBank mRNA BC072714.1 - 3 - 4 502 103
BC165782.1 - 3 - 4 517 103

View Full Product Details