Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spag6
Entrez Gene ID
431757
Gene Name
sperm associated antigen 6
Gene Aliases
zgc:91805
Chromosome Location
Chr.24: 17269872 - 17276771 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002210.1 NP_001002210.1 10 - 11 1568 118
GenBank mRNA BC074072.1 - 10 - 11 1568 118
BC165187.1 - 10 - 11 1488 118

View Full Product Details