Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mterf3
Entrez Gene ID
436888
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
mterfd1, zgc:92270
Chromosome Location
Chr.19: 44058557 - 44065557 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002615.1 NP_001002615.1 3 - 4 522 77
GenBank mRNA BC075975.1 - 3 - 4 522 77

View Full Product Details