Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
calcrla
Entrez Gene ID
445508
Gene Name
calcitonin receptor-like a
Gene Aliases
im:7141310, zgc:100872
Chromosome Location
Chr.9: 42360679 - 42418466 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004010.1 NP_001004010.1 11 - 12 1364 136
GenBank mRNA BC080226.1 - 11 - 12 1364 136
BC129300.1 - 11 - 12 1366 136

View Full Product Details