Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
quo
Entrez Gene ID
321031
Gene Name
quattro
Gene Aliases
cb9, fb97g10, sb:cb9, wu:fb97g10
Chromosome Location
Chr.6: 2030703 - 2092501 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004109.2 NP_001004109.2 5 - 6 2615 78
GenBank mRNA AY654285.2 - 5 - 6 2614 78
BC162633.1 - 5 - 6 2535 78

View Full Product Details