Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atad1a
Entrez Gene ID
368672
Gene Name
ATPase family AAA domain containing 1a
Gene Aliases
ATPase, si:zc156n3.1, zgc:101570
Chromosome Location
Chr.5: 16718919 - 16734062 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004640.3 NP_001004640.1 2 - 3 310 81
GenBank mRNA BC081379.1 - 2 - 3 309 81
BC164811.1 - 1 - 2 206 81
EH554125.1 - 2 - 3 345 81

View Full Product Details