Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atad1a
Entrez Gene ID
368672
Gene Name
ATPase family AAA domain containing 1a
Gene Aliases
ATPase, si:zc156n3.1, zgc:101570
Chromosome Location
Chr.5: 16718919 - 16734062 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004640.3 NP_001004640.1 6 - 7 832 66
GenBank mRNA BC081379.1 - 6 - 7 66
BC164811.1 - 5 - 6 66
EH565751.1 - 4 - 5 657 66

View Full Product Details