Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
parapinopsinb
Entrez Gene ID
368725
Gene Name
parapinopsin b
Gene Aliases
opsin, pp2, si:busm1-136d19.2, si:dz136d19.2, vispp
Chromosome Location
Chr.11: 36777613 - 36785073 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005312.1 NP_001005312.1 1 - 2 343 74
GenBank mRNA AB626967.1 - 1 - 2 400 74
KT008405.1 - 1 - 2 343 74

View Full Product Details