Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klhl6
Entrez Gene ID
368818
Gene Name
kelch-like family member 6
Gene Aliases
si:dz198m22.2, zgc:152803
Chromosome Location
Chr.2: 36646499 - 36659156 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005316.2 NP_001005316.2 4 - 5 1163 67

View Full Product Details