Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sccpdhb
Entrez Gene ID
407665
Gene Name
saccharopine dehydrogenase b
Gene Aliases
cb4, sb:cb4, zgc:101527
Chromosome Location
Chr.20: 34967423 - 34985555 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005574.1 NP_001005574.1 1 - 2 401 99
GenBank mRNA BC066701.1 - 1 - 2 186 99
BC083193.1 - 1 - 2 401 99

View Full Product Details