Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tlcd1
Entrez Gene ID
450051
Gene Name
TLC domain containing 1
Gene Aliases
zgc:101692
Chromosome Location
Chr.21: 26080215 - 26089899 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006071.1 NP_001006071.1 2 - 3 366 77
GenBank mRNA BC083250.1 - 2 - 3 366 77

View Full Product Details