Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crygm5
Entrez Gene ID
474328
Gene Name
crystallin, gamma M5
Gene Aliases
zgc:103622
Chromosome Location
Chr.9: 162106 - 163440 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007058.2 NP_001007059.1 2 - 3 274 133
GenBank mRNA AY738754.1 - 1 - 2 259 133
BC083443.1 - 2 - 3 275 133
BC164872.1 - 1 - 2 288 133

View Full Product Details