Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lifra
Entrez Gene ID
541493
Gene Name
LIF receptor subunit alpha a
Gene Aliases
lifr.a, zgc:112989
Chromosome Location
Chr.5: 8417184 - 8483537 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014306.2 NP_001014328.2 9 - 10 1416 103
GenBank mRNA BC091862.1 - 9 - 10 1420 103
BC165177.1 - 8 - 9 1246 103

View Full Product Details