Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pnocb
Entrez Gene ID
394236
Gene Name
prepronociceptin b
Gene Aliases
pnoc, zgc:136385
Chromosome Location
Chr.20: 34717514 - 34734002 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015044.1 NP_001015044.1 2 - 3 255 71
GenBank mRNA BC115105.1 - 2 - 3 262 71

View Full Product Details