Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
nadka
Entrez Gene ID
550242
Gene Name
NAD kinase a
Gene Aliases
zgc:110083
Chromosome Location
Chr.8: 53051729 - 53069540 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017580.1 NP_001017580.1 - 1266 75
GenBank mRNA BC092723.1 - - 1266 75
BC165884.1 - - 1116 75

View Full Product Details