The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017580.1 | NP_001017580.1 | - | 1013 | 75 |
GenBank mRNA | BC092723.1 | - | - | 1013 | 75 |
BC165884.1 | - | - | 863 | 75 |