Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acer1
Entrez Gene ID
550266
Gene Name
alkaline ceramidase 1
Gene Aliases
asah3, zgc:110285
Chromosome Location
Chr.8: 20232257 - 20246014 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017603.1 NP_001017603.1 2 - 3 274 108
XM_005166840.4 XP_005166897.1 2 - 3 396 108
GenBank mRNA BC092849.1 - 2 - 3 274 108
BC164606.1 - 1 - 2 128 108

View Full Product Details