Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efcab11
Entrez Gene ID
550510
Gene Name
EF-hand calcium binding domain 11
Gene Aliases
zgc:110317
Chromosome Location
Chr.17: 16423721 - 16533069 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017812.1 NP_001017812.1 4 - 5 419 77
XM_021467159.1 XP_021322834.1 3 - 4 387 77
GenBank mRNA BC092868.1 - 4 - 5 419 77

View Full Product Details