Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ovca2
Entrez Gene ID
553576
Gene Name
OVCA2 serine hydrolase domain containing
Gene Aliases
zgc:110011
Chromosome Location
Chr.10: 35664129 - 35668579 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020555.1 NP_001018391.1 1 - 2 383 78
GenBank mRNA BC095129.1 - 1 - 2 383 78

View Full Product Details