Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
il22
Entrez Gene ID
553964
Gene Name
interleukin 22
Gene Aliases
-
Chromosome Location
Chr.4: 13544595 - 13548806 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020792.1 NP_001018628.1 3 - 4 488 60
GenBank mRNA BC163192.1 - 2 - 3 743 60

View Full Product Details