Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hivep2a
Entrez Gene ID
562048
Gene Name
HIVEP zinc finger 2a
Gene Aliases
hivep2, si:ch211-157m21.1
Chromosome Location
Chr.20: 37511450 - 37636368 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030164.1 NP_001025335.1 6 - 7 6889 149

View Full Product Details