Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sult4a1
Entrez Gene ID
678517
Gene Name
sulfotransferase family 4A, member 1
Gene Aliases
zgc:136828
Chromosome Location
Chr.4: 8797114 - 8810740 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040244.1 NP_001035334.1 2 - 3 386 91
GenBank mRNA BC115282.1 - 2 - 3 386 91

View Full Product Details