Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wdyhv1
Entrez Gene ID
560935
Gene Name
WDYHV motif containing 1
Gene Aliases
ntaq1, si:rp71-45k5.3
Chromosome Location
Chr.19: 35002479 - 35004947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044947.1 NP_001038412.1 4 - 5 382 64
GenBank mRNA BC171472.1 - 4 - 5 436 64
BC171474.1 - 4 - 5 436 64

View Full Product Details