Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glsa
Entrez Gene ID
564147
Gene Name
glutaminase a
Gene Aliases
glsl
Chromosome Location
Chr.22: 12826282 - 12862484 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045044.1 NP_001038509.1 6 - 7 828 71
XM_005161685.4 XP_005161742.1 6 - 7 890 71
XM_009296204.3 XP_009294479.1 6 - 7 897 71

View Full Product Details