Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mthfsd
Entrez Gene ID
566203
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
fc43c12, si:dkey-199i24.1, wu:fc43c12, zgc:153374
Chromosome Location
Chr.18: 30878075 - 30949716 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045099.1 NP_001038564.1 2 - 3 236 146
GenBank mRNA BC122258.1 - 2 - 3 236 146
BC171684.1 - 2 - 3 153 146
BC171688.1 - 2 - 3 152 146

View Full Product Details