Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptpreb
Entrez Gene ID
569164
Gene Name
protein tyrosine phosphatase receptor type Eb
Gene Aliases
si:dkey-46n18.4
Chromosome Location
Chr.17: 29319544 - 29343657 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045177.1 NP_001038642.1 14 - 15 1507 93
XM_005158789.3 XP_005158846.1 14 - 15 1559 93
XM_017351664.2 XP_017207153.1 11 - 12 1248 93
GenBank mRNA BC128883.1 - 14 - 15 1518 93
FN653011.1 - 1 - 2 111 93

View Full Product Details