Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptpreb
Entrez Gene ID
569164
Gene Name
protein tyrosine phosphatase receptor type Eb
Gene Aliases
si:dkey-46n18.4
Chromosome Location
Chr.17: 29319544 - 29343657 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045177.1 NP_001038642.1 4 - 5 445 91
XM_005158789.3 XP_005158846.1 4 - 5 488 91
GenBank mRNA BC128883.1 - 4 - 5 446 91

View Full Product Details