Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
neurl1aa
Entrez Gene ID
561943
Gene Name
neuralized E3 ubiquitin protein ligase 1Aa
Gene Aliases
neurl, neurl1a, neurla, si:dkey-82d4.1, si:dkey-82d4.3, zgc:153971
Chromosome Location
Chr.1: 47701932 - 47828227 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077558.2 NP_001071026.1 4 - 5 1434 81
XM_021475292.1 XP_021330967.1 4 - 5 1551 81
GenBank mRNA BC125926.1 - 4 - 5 1434 81

View Full Product Details