Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
syt10
Entrez Gene ID
564226
Gene Name
synaptotagmin X
Gene Aliases
si:ch211-231h23.1, si:dkey-207j16.1
Chromosome Location
Chr.4: 234501 - 251337 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082842.1 NP_001076311.1 1 - 2 147 68

View Full Product Details