Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vps72a
Entrez Gene ID
100001285
Gene Name
vacuolar protein sorting 72 homolog a
Gene Aliases
si:dkeyp-24a7.1, vps72
Chromosome Location
Chr.19: 366225 - 371568 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082944.1 NP_001076413.1 5 - 6 708 116

View Full Product Details