Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
usp12a
Entrez Gene ID
565736
Gene Name
ubiquitin specific peptidase 12a
Gene Aliases
sb:eu882, usp12, wu:fi09d12
Chromosome Location
Chr.24: 21600320 - 21620275 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083556.1 NP_001077025.1 2 - 3 154 62
GenBank mRNA BC115286.1 - 2 - 3 154 62

View Full Product Details