Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il15ra
Entrez Gene ID
100192216
Gene Name
interleukin 15 receptor subunit alpha
Gene Aliases
il-15ra, zgc:195175
Chromosome Location
Chr.4: 32525 - 50390 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083584.2 NP_001077053.1 6 - 7 684 124
NM_001359926.1 NP_001346855.1 5 - 6 666 124
GenBank mRNA EF143578.1 - 6 - 7 684 124
EF165721.1 - 5 - 6 666 124

View Full Product Details