Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dync2i2
Entrez Gene ID
794361
Gene Name
dynein 2 intermediate chain 2
Gene Aliases
dync2i1, wdr34, zgc:171284
Chromosome Location
Chr.21: 13216906 - 13225421 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105690.1 NP_001099160.1 4 - 5 929 85
XM_021468923.1 XP_021324598.1 4 - 5 948 85
GenBank mRNA BC095866.1 - 4 - 5 927 85
BC133909.1 - 4 - 5 534 85
BC152526.1 - 4 - 5 929 85

View Full Product Details