Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2aa12
Entrez Gene ID
555510
Gene Name
cytochrome P450, family 2, subfamily AA, polypeptide 12
Gene Aliases
zgc:171945
Chromosome Location
Chr.23: 42292748 - 42306332 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109701.2 NP_001103171.2 2 - 3 383 74
GenBank mRNA BC152006.1 - 2 - 3 383 74

View Full Product Details