Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atp1a2a
Entrez Gene ID
64609
Gene Name
ATPase Na+/K+ transporting subunit alpha 2a
Gene Aliases
atp1a2, atp[a]2, atpa2, cb702, fa10d04, fa10d05, wu:fa10d05
Chromosome Location
Chr.2: 111938 - 172731 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131683.1 NP_571758.1 6 - 7 725 87
GenBank mRNA AF286373.1 - 6 - 7 725 87
AY028628.1 - 6 - 7 659 87
BC163629.1 - 6 - 7 647 87

View Full Product Details