Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ddi2
Entrez Gene ID
386644
Gene Name
DNA-damage inducible protein 2
Gene Aliases
zgc:63515
Chromosome Location
Chr.6: 40554492 - 40563510 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198375.2 NP_938189.2 4 - 5 751 86
XM_005166024.4 XP_005166081.1 4 - 5 890 86
XM_021476799.1 XP_021332474.1 4 - 5 889 86
GenBank mRNA AY398334.1 - 4 - 5 772 86
BC055129.1 - 4 - 5 751 86

View Full Product Details