Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ddi2
Entrez Gene ID
386644
Gene Name
DNA-damage inducible protein 2
Gene Aliases
zgc:63515
Chromosome Location
Chr.6: 40554492 - 40563510 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198375.2 NP_938189.2 5 - 6 879 76
XM_005166024.4 XP_005166081.1 5 - 6 1018 76
XM_021476799.1 XP_021332474.1 5 - 6 1017 76
GenBank mRNA BC055129.1 - 5 - 6 879 76

View Full Product Details